Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C4598

Sigma-Aldrich

Anti-Chapsyn-110 antibody produced in rabbit

purified immunoglobulin, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot (chemiluminescent): 1:300-1:1,000

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Dlg2(64053)

Immunogen

highly purified fusion protein of Schistosoma japonicum glutathione-S-transferase (GST) corresponding to residues 336-379 of rat chapsyn-110.

Physical form

Lyophilized at 1.0 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Preparation Note

Purified using Protein A

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service