Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C4491

Sigma-Aldrich

Anti-Calcium-Sensing Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CASR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4-8 μg/mL using human parathroid gland tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CASR(846)
mouse ... Casr(12374)
rat ... Casr(24247)

General description

The CASR (calcium sensing receptor) gene is mapped to human chromosome 3q13.33-q21.1. It is ubiquitously expressed and codes for protein containing 1,078 amino acid residues. The protein structure is divided into three domains : a large, extracellular domain at the amino terminal, a transmembrane domain and an intracellular carboxyl domain.

Immunogen

synthetic peptide corresponding to the extracellular N-terminal domain of human calcium-sensing receptor, conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Application

Anti-calcium-sensing receptor antibody produced in rabbit is suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 4-8 μg/mL using human parathyroid gland tissue.

Biochem/physiol Actions

CASR (calcium sensing receptor) is an integral membrane protein. It senses changes in the extracellular concentration of calcium ions and maintains its homeostasis within the system. The activity of this receptor is mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system. The antibody specifically recognizes human calcium sensing receptor by immunohistochemistry with formalin-fixed, paraffin-embedded tissues. Calcium sensing receptor is actively involved in the secretion of peptide hormones such as parathyroid hormone, corticotropin hormone, adrenocorticotropic hormone, gastrin, insulin and growth hormone. It is associated with the function of K+, Ca2+ and water ion channels. Regulation of pituitary tumor-transforming gene expression is controlled by CasR. CasR is associated with cell proliferation (colonic/ovarian surface epithelial cells, testis Leydig cell cancer cells, fibroblasts, and keratinocytes) and cell differentiation (keratinocytes, goblet cells and mammary epithelial cells) processes. It mediates chemotaxis of pre osteoblastic cells and macrophages. It regulates apoptosis in cancer cells including Leydig cell tumour and prostate cancer cell line.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The calcium-sensing receptor in normal physiology and pathophysiology: a review.
Tfelt-Hansen J and Brown E M
Critical Reviews in Clinical Laboratory Sciences, 42(1), 35-70 (2005)
The emerging role of genomics in the diagnosis and workup of congenital urinary tract defects: a novel deletion syndrome on chromosome 3q13. 31-22.1.
Materna-Kiryluk A, et al.
Pediatric Nephrology (Berlin), 29(2), 257-267 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service