Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C433060

Sigma-Aldrich

DMT-dC(ac)-CPG Column

pore size 1000 Å, 1 μmol, configured for ABI, configured for PerkinElmer 8900

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

product line

Proligo Reagents

pore size

1000 Å

compatibility

configured for ABI
configured for PerkinElmer 8900

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service