Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C421060

Sigma-Aldrich

DMT-dC(bz)-CPG column

pore size 1000 Å, 0.2 μmol, configured for PerkinElmer, configured for ABI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

type

for DNA synthesis

product line

Proligo Reagents

assay

≥95.0% (Nucleoside Purity, reversed phase HPLC)

pore size

1000 Å

capacity

≥99 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI
configured for PerkinElmer

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service