Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-41222

Sigma-Aldrich

DetachKit 2

3 x 250 ml

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 250 ML
$212.00

$212.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 250 ML
$212.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$212.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 3 × 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

cooler

storage temp.

−20°C

General description

DetachKit 2-250

Application

The DetachKit 2 was developed for gentle and effective detachment of adherent primary human cells in culture and is especially valuable for more sensitive cell types. Each batch is tested on primary human cells.

Components

Each DetachKit 2 consists of three components: HEPES BSS (HEPES buffered Balanced Salt Solution), Trypsin/EDTA Solution and TNS (Trypsin Neutralization Solution). The DetachKit 2 comes with a Lower Trypsin/EDTA ratio of 0.025%/0.01%. HEPES BSS contains 30 mM HEPES, D-Glucose, NaCl, KCl, Na-Phosphate and Phenol Red. TNS contains 0.05% Trypsin Inhibitor from soybean and 0.1% Bovine Serum Albumin.

Quality

All lots of PromoCell′s DetachKits are subjected to comprehensive quality control tests. Each lot is routinely tested for use with primary human cells, the absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store at -20°C in the dark immediately after arrival. Refrigerate thawed solution at 4 to 8°C. If stored properly, the product is stable until the expiry date stated on the label. Thawed solutions can be stored at 4 to 8°C and used for up to 6 weeks.

Subculture Routine

Click here for more information.

pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - STOT RE 2

target_organs

Respiratory Tract

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service