C3896
CM-Cellulose
fibers
Synonym(s):
Carboxymethyl cellulose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
fibers
capacity
~0.6 meq/g
InChI
1S/C6H12O6.C2H4O2/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4/h1,3-6,8-12H,2H2;1H3,(H,3,4)
Inchi Key
VJHCJDRQFCCTHL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
CM Cellulose products are used in protein chromatography, ion exchange chromatography and cation exchange media. CM Cellulose has been used in studies to inform the suppressed proliferation of hepatoma and leukemic cells and the inhibition of the human immunodeficiency virus type 1 (HIV-1).
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yingying Wu et al.
The Journal of antibiotics, 65(2), 67-72 (2011-12-22)
A 14.6-kDa RNase, with a pH optimum of 5.5 and a temperature optimum of 70 °C, was isolated from dried fruiting bodies of the edible mushroom Lactarius flavidulus. The purification procedure involved, in succession, ion exchange chromatography on DEAE-cellulose, CM-cellulose and
Yingying Wu et al.
Carbohydrate research, 346(16), 2576-2581 (2011-10-04)
The isolation of a dimeric 29.8-kDa lectin (LFL) from dried Lactarius flavidulus fruit bodies is reported herein. The chromatographic procedure utilized comprised anion-exchange chromatography on DEAE-cellulose, cation-exchange chromatography on CM-cellulose, anion-exchange chromatography on SP-Sepharose, and gel filtration by fast protein
T Subramani et al.
The protein journal, 31(2), 120-128 (2011-12-17)
Carboxylesterases are hydrolases which catalyze the hydrolysis of various types of esters. Carboxylesterase from the seeds of Jatropha curcas has been purified to homogeneity using ammonium sulfate fractionation, CM-cellulose chromatography, Sephadex G-100 chromatography and preparative polyacrylamide gel electrophoresis (PAGE). The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service