C3835
Ciliary Neurotrophic Factor from rat
recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE), carrier free
Synonym(s):
CNTF
About This Item
Recommended Products
biological source
rat
recombinant
expressed in E. coli
assay
≥98% (SDS-PAGE)
form
lyophilized powder
mol wt
22.7 kDa
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
color
white
solubility
water: soluble 0.10 mL, clear to slightly hazy, colorless
UniProt accession no.
storage temp.
−20°C
Gene Information
rat ... Cntf(25707)
Biochem/physiol Actions
Physical form
Analysis Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service