Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3615

Sigma-Aldrich

Control primer set for Arabidopsis Actin-2 gene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

storage temp.

−20°C

General description

This product consists of a primer set for the Arabidopsis housekeeping gene Actin-2 (At3g18780). It is intended for use as an internal control for Q-RT-PCR for expression analysis of any Arabidopsis gene.

Of our three control primer series for Arabidopsis gene expression, Actin-2 control primer is designed as a control for high-abundant gene expression. The other two control primers in the series are nuclear cap-binding protein (CBP20, Product Code C3115) and ubiquitin (UBC, Product Code C3240) designed as controls for medium- to low-abundant gene expression.

Components

Each tube contains enough primer (forward or reverse) for 100 RT-PCR reactions (50 μl reaction volume)
  • Forward Primer (Tm = 68 °C based on % GC method in 1 M salt concentration), 20 μM, in TE buffer, Product Code A7728.
  • Reverse Primer (Tm = 67 °C based on % GC method in 1 M salt concentration), 20 μM, in TE buffer, Product Code A6978.

Suitability

Primer should be stored at -20 °C. Primers are stable at -20 °C for at least 1 year and can be frozen and thawed at least 10 times.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service