Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3606

Greiner CapMat for sealing multiwell plates

96 well CapMat, for sealing 96 well plates, round nap, translucent, non-pierceable

Synonym(s):

multiwell plate cap mats, Greiner CapMats for sealing multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.22

material

ethylene vinyl acetate

sterility

non-sterile

packaging

case of 50 ea (internal packs of 10)

manufacturer/tradename

Greiner 381070

size

96 wells

color

translucent

suitability

suitable for (sealing 96 well plates)

Looking for similar products? Visit Product Comparison Guide

Chem/Phys Resistance

Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service