C3421
Worthington Accessories
stainless steel canister for 35 L Dewar
All Photos(1)
About This Item
UNSPSC Code:
41103000
Recommended Products
manufacturer/tradename
Taylor-Wharton
size
3.7 in. (9.4 cm) × 11 in. (27.9 cm)
General description
For storage and easy retrieval of vials in canes (canes not included).
Choose from one of the most recent versions:
Certificates of Analysis (COA)