Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3277

Sigma-Aldrich

Cedar oil

for microscopy

Synonym(s):

Cedarwood oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211

refractive index

n20/D ~1.519 (lit.)

density

0.952 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Immersion oil for microscopy

Biochem/physiol Actions

Cedar oil demonstrates antimicrobial activity against anaerobic bacteria and yeast, as assessed by a standardized broth assay [1].

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanfeng Liu et al.
Oncology letters, 10(2), 1184-1190 (2015-12-02)
The aim of the present study was to investigate the long-term survival of patients with refractory acute promyelocytic leukemia (APL) that were administered alternately with compound realgar natural indigo tablet (CRNIT) treatment and chemotherapy. In total, 31 patients with refractory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service