Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3239

Sigma-Aldrich

Anti-Calpain-94 (Catalytic Domain), Large Subunit antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAPN3(825)

Specificity

Detects all isoforms of calpain 94. Recognizes latent and active calpain 94.

Immunogen

synthetic peptide corresponding to the catalytic domain of the large subunit of human calpain 94 (calpain 3, nCANP, nCL-1, calpain p94, muscle calpain).

Physical form

Solution is 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailee R Perrett et al.
Cell reports, 42(5), 112524-112524 (2023-05-20)
Lassa fever is an acute hemorrhagic fever caused by the zoonotic Lassa virus (LASV). The LASV glycoprotein complex (GPC) mediates viral entry and is the sole target for neutralizing antibodies. Immunogen design is complicated by the metastable nature of recombinant
Bispecific antibodies combine breadth, potency, and avidity of parental antibodies to neutralize sarbecoviruses.
RadiA?, et al.
iScience, 26, 106540-106540 (2023)
Mitch Brinkkemper et al.
iScience, 27(5), 109719-109719 (2024-05-06)
Zoonotic events by sarbecoviruses have sparked an epidemic (severe acute respiratory syndrome coronavirus [SARS-CoV]) and a pandemic (SARS-CoV-2) in the past two decades. The continued risk of spillovers from animals to humans is an ongoing threat to global health and
Philip J M Brouwer et al.
Cell host & microbe, 30(12), 1759-1772 (2022-11-19)
The Lassa virus is endemic in parts of West Africa, and it causes hemorrhagic fever with high mortality. The development of a recombinant protein vaccine has been hampered by the instability of soluble Lassa virus glycoprotein complex (GPC) trimers, which
Mitch Brinkkemper et al.
iScience, 25(12), 105649-105649 (2022-11-29)
The emergence of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) variants poses continuous challenges in combating the virus. Here, we describe vaccination strategies to broaden SARS-CoV-2 and sarbecovirus immunity by combining spike proteins based on different viruses or viral strains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service