Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3022

Sigma-Aldrich

Z-Gly-Gly-Leu p-nitroanilide

protease substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N5O7
CAS Number:
Molecular Weight:
499.52
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

assay

≥98% (TLC)

form

powder

solubility

methanol: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)CNC(=O)CNC(=O)OCc1ccccc1)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C24H29N5O7/c1-16(2)12-20(23(32)27-18-8-10-19(11-9-18)29(34)35)28-22(31)14-25-21(30)13-26-24(33)36-15-17-6-4-3-5-7-17/h3-11,16,20H,12-15H2,1-2H3,(H,25,30)(H,26,33)(H,27,32)(H,28,31)

InChI key

IHRYETONKBXGOF-UHFFFAOYSA-N

Substrates

A sensitive chromogenic substrate for subtilisins and neutral endopeptidases.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new chromogenic substrate for subtilisin.
L A Lyublinskaya et al.
Analytical biochemistry, 62(2), 371-376 (1974-12-01)
Degradation of bradykinin by isolated neutral endopeptidases of brain and pituitary.
S Wilk et al.
Biochemical and biophysical research communications, 90(1), 1-6 (1979-09-12)
Ajay Kumar Shaw et al.
Journal of photochemistry and photobiology. B, Biology, 86(3), 199-206 (2006-11-18)
Enzymatic activity of a proteolytic enzyme Subtilisin Carlsberg (SC) in anionic sodium dodecyl sulfate (SDS) micellar medium has been explored and found to be retarded compared to that in bulk buffer. Circular dichroism (CD) study reveals that SDS, which is
S Wilk et al.
Journal of neurochemistry, 40(3), 842-849 (1983-03-01)
Pituitary cation-sensitive neutral endopeptidase splits peptide bonds on the carboxyl side of hydrophobic amino acids (chymotrypsin-like activity), basic amino acids (trypsin-like activity), and acidic amino acids (peptidyl-glutamyl-peptide bond hydrolyzing activity). All three activities copurify, are inhibited by cations, and reside
B J Wagner et al.
Experimental eye research, 38(5), 477-483 (1984-05-01)
Lens neutral proteinase is thought to exhibit primarily endopeptidase activity. We have identified a synthetic endopeptidase substrate which is hydrolyzed by the bovine lens neutral proteinase preparation. Among 11 fluoro- and chromogenic endopeptidase substrates, only carbobenzoxy-glycylglycyl-L-leucyl-p-nitroanilide is effectively hydrolyzed. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service