PromoCell’s human pericytes derived from microvessels of the human placenta are cryopreserved at the end of secondary culture (P1) and after thawing are considered P2. The number of populations doublings is not determined for each individual cell lot, however they are guarenteed to be grown for at least 15 population doublings.
C-28040
Pericyte Growth Medium
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
About This Item
Recommended Products
form
aqueous solution
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Quality
Storage and Stability
Subculture Routine
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
What are pericytes? What are common pericyte markers? Are pericytes mesenchymal stem cells? Learn more about pericytes in our FAQ page. Explore over 350 PromoCell products.
Primary human pericytes and cell culture media optimized for the in vitro cultivation of human pericytes from all tissues. Protocols for handling, cell passaging, media, and product use.
-
How long do pericytes grow in culture?
1 answer-
Helpful?
-
-
What markers are used to characterize pericytes?
1 answer-
Pericytes are positive for CD146, PDGFRβ, αSMA, CD13, NG2, Desmin, and negative for CD31, CD34, CD45, and CD56.
Helpful?
-
-
How are pericytes isolated?
1 answer-
Pericyte isolation generally starts with enzymatic digestion of tissue with Trypsin EDTA, usually followed by the isolation of microvessel fragments via successive filtration steps. Pure pericyte cultures can be obtained through either positive or negative immuno-selection via the use of magnetic beads or flow cytometry using the antibody markers above.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service