C212030
DMT-2′O-TBDMS-rC(tac) Phosphoramidite
configured for ABI
Synonym(s):
DMT-2′-O-TBDMS-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-?methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]
About This Item
product line
Proligo Reagents
assay
≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)
form
solid
technique(s)
oligo synthesis: suitable
λ
conforms (UV/VIS Identity)
compatibility
configured for ABI
nucleoside profile
base: cytidine
base protecting group: TAC
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast
storage temp.
−20°C
SMILES string
COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(=O)COc4ccc(cc4)C(C)(C)C)=NC3=O)(c5ccccc5)c6ccc(OC)cc6
InChI
1S/C57H76N5O10PSi/c1-39(2)62(40(3)4)73(69-36-18-34-58)71-51-48(37-68-57(42-19-16-15-17-20-42,43-23-27-45(65-11)28-24-43)44-25-29-46(66-12)30-26-44)70-53(52(51)72-74(13,14)56(8,9)10)61-35-33-49(60-54(61)64)59-50(63)38-67-47-31-21-41(22-32-47)55(5,6)7/h15-17,19-33,35,39-40,48,51-53H,18,36-38H2,1-14H3,(H,59,60,63,64)/t48-,51-,52-,53-,73?/m1/s1
InChI key
FAODNIJMAKFTFL-WSFKJFENSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service