Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C21200-HH

Sigma-Aldrich

DMT-2′O-TBDMS-rC(tac) Phosphoramidite

Synonym(s):

DMT-2′-O-TBDMS-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-?methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H76N5O10PSi
CAS Number:
Molecular Weight:
1050.30
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Proligo Reagents

assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

solid

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

nucleoside profile

base: cytidine
base protecting group: TAC
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(=O)COc4ccc(cc4)C(C)(C)C)=NC3=O)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C57H76N5O10PSi/c1-39(2)62(40(3)4)73(69-36-18-34-58)71-51-48(37-68-57(42-19-16-15-17-20-42,43-23-27-45(65-11)28-24-43)44-25-29-46(66-12)30-26-44)70-53(52(51)72-74(13,14)56(8,9)10)61-35-33-49(60-54(61)64)59-50(63)38-67-47-31-21-41(22-32-47)55(5,6)7/h15-17,19-33,35,39-40,48,51-53H,18,36-38H2,1-14H3,(H,59,60,63,64)/t48-,51-,52-,53-,73?/m1/s1

InChI key

FAODNIJMAKFTFL-WSFKJFENSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service