Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1989

Sigma-Aldrich

Anti-Calpain-9 (Domain I), Large Subunit antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAPN9(10753)
mouse ... Capn9(73647)
rat ... Capn9(116694)

Immunogen

synthetic peptide corresponding to the amino-terminal end of domain I of the large subunit of human calpain 9 (capn-9, nCL-4, GC36).

Physical form

Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service