C1897
Catechol-O-methyl Transferase from porcine liver
lyophilized powder, ≥150 units/mg protein
Synonym(s):
COMT, Pyrocatechol-O-methyl Transferase, S-Adenosyl-L-methionine:catechol O-methyltransferase
About This Item
Recommended Products
form
lyophilized powder
specific activity
≥150 units/mg protein
purified by
chromatography
composition
Protein, 30-60% biuret
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Unit Definition
Previous radioactive assay unit definition: One unit will catalyze the methylation of 1.0 nanomole of protocatechuic acid per hr at pH 7.9 at 37°C using S-adenosyl-L-[methyl-14C]methionine as the methyl donor. (~6 radioactive units equal one new unit).
Physical form
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service