Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1867

Sigma-Aldrich

Carrageenan

Non-gelling, Mixture of κ and λ carrageenans

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201

biological source

algae (marine)

form

powder

quality

Non-gelling

Looking for similar products? Visit Product Comparison Guide

General description

Carrageenans are mucopolysaccharides from the cell walls of the red algae. They are anionic linear polymers composed of 1,3α-1,4β-galactans having one (κ-), two (ι-) or three (λ-) sulfates per disaccharide unit. In ionic solutions, κ- and ι-carrageenans self-associate into helical structures that form rigid or flexible gels, respectively. λ-carrageenans do not form helices and are non-gelling. Carrageenans are used commercially as thickeners and stabilizing agents.

Application

Carrageenans are used to suppress immune response in vivo and in vitro via mechanisms believed to involve selective cytopathic effect on macrophages. As ι-, κ-, and λ-carrageenans have varying degress of sulfation, they may be studied comparatively for differences in protective, antiviral, and immunosuppression activities.
Non-gelling carrageenan is used to induce inflammation and inflammatory pain in the rodent hindpaw or air pouch models. More recently carrageenan has been injected directly into the joints of rodents to model arthritic pain and inflammation.

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A F Gabriel et al.
Journal of neuroscience methods, 163(1), 9-16 (2007-03-27)
Experimental pain research is often complicated by the absence of an objective and detailed method to analyze behavioral changes. In the present study, acute pain was induced into the right knee of the rat (n=15) through the injection of 2mg
A D Sedgwick et al.
Agents and actions, 18(3-4), 439-446 (1986-06-01)
A comparative assessment of the time course of changes in three models of acute carrageenan-induced inflammation, the six-day air pouch, polyester sponge and pleurisy models, was obtained by measuring exudate volume, leucocyte numbers, PGE2 concentrations and lactate dehydrogenase activity at
A W Thomson et al.
Agents and actions, 11(3), 265-273 (1981-05-01)
Carrageenans (kappa, lambda and iota) are sulphated polysaccharides isolated from marine algae that can markedly suppress immune responses both in vivo and in vitro. Impairment of complement activity and humoral responses to T-dependent antigens, depression of cell-mediated immunity, prolongation of
M L Weiner
Agents and actions, 32(1-2), 46-51 (1991-01-01)
Food-grade carrageenan is a safe natural product prepared from seaweed. Its addition to food imparts many desirable characteristics which have allowed it to be used continuously for centuries. The long safe history of this natural food additive is confirmed by
S Nicklin et al.
Food additives and contaminants, 6(4), 425-436 (1989-10-01)
Carrageenans are a group of high molecular weight sulphated polygalactans which find extensive use in the food industry as thickening, gelling and protein-suspending agents. Although there is no evidence to suggest that the persorption of small amounts of carrageenans across

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service