Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1845

Sigma-Aldrich

Cofilin from chick embryo

≥90%, recombinant, expressed in E. coli, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

recombinant

expressed in E. coli

assay

≥90%

form

lyophilized powder

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... CFL2(423320)

Biochem/physiol Actions

Low molecular mass protein which modulates the binding of actin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Renoult et al.
The Journal of biological chemistry, 274(41), 28893-28899 (1999-10-03)
The cofilins are members of a protein family that binds monomeric and filamentous actin, severs actin filaments, and increases monomer off-rate from the pointed end. Here, we characterize the cofilin-actin interface. We confirm earlier work suggesting the importance of the
H Aizawa et al.
Genes to cells : devoted to molecular & cellular mechanisms, 4(6), 311-324 (1999-07-28)
Cofilin is a low-molecular weight actin-modulating protein, which binds to, severs, and depolymerizes actin filaments in vitro. Aip1, an actin-interacting protein, was recently identified as a product of a gene on a multicopy plasmid which suppresses the temperature-sensitive phenotype of
Nicolas Sémiramoth et al.
Journal of leukocyte biology, 85(2), 310-321 (2008-11-19)
The innate immune response to enteropathogenic bacteria includes chemokine-induced polymorphonuclear neutrophil (PMN) migration across mucosal epithelia leading to bacterial clearance and resolution of infection. Among these bacteria, diffusely adherent Escherichia coli expressing Afa/Dr fimbriae (Afa/Dr DAEC), causing childhood diarrhea, can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service