Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C176

Sigma-Aldrich

CGP-23996

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

form

solid

Quality Level

mol wt

1474 (peptide free base)

color

white

solubility

0.1 M acetic acid: soluble

shipped in

dry ice

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)C[C@@H]3NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCCCC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC3=O)CCCCN)[C@H](O)C)Cc6ccc(cc6)O)[C@H](O)C)CO)C(=O)O)CCCCN)CC(=O)N)Cc5ccccc5)Cc4ccccc4)cccc2

InChI

1S/C73H99N15O18/c1-41(90)61-71(103)85-55(36-45-28-30-47(92)31-29-45)69(101)88-62(42(2)91)72(104)86-58(40-89)70(102)80-52(73(105)106)26-10-5-11-27-60(94)78-50(24-14-16-32-74)63(95)84-57(38-59(76)93)68(100)82-53(34-43-18-6-3-7-19-43)65(97)81-54(35-44-20-8-4

InChI key

HTEPNQQGQXAYEW-MTBXKZQJSA-N

Application

Non-reducible analog of somatostatin suitable for iodination. The radio-iodinated molecule can be used in binding assays for somatostatin receptors.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Raynor et al.
Journal of neurochemistry, 59(4), 1241-1250 (1992-10-01)
Somatostatin (SRIF) is a neurotransmitter that produces its multiple effects in the CNS through interactions with membrane-bound receptors. Subtypes of SRIF receptors are found in the CNS that are distinguished by their sensitivities to the cyclic hexapeptide MK-678, such that
K Raynor et al.
The Journal of pharmacology and experimental therapeutics, 264(1), 110-116 (1993-01-01)
GH3 cells express receptors for the neuropeptide somatostatin (SRIF). In the present study, we have identified and characterized SRIF1 and SRIF2 receptors in GH3 cells using the radioligands [125I]MK 678 and [125I]CGP 23996. [125I]MK 678 binding to SRIF1 receptors was
J L Martin et al.
Neuroscience, 41(2-3), 581-593 (1991-01-01)
Somatostatin receptor subtypes were labeled with the somatostatin analogs [125I]CGP 23996 and [125I]MK 678 and the distribution of these receptors in rat brain was investigated using quantitative autoradiographic techniques. [125I]CGP 23996 and [125I]MK 678 specifically label different populations of somatostatin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service