Key Documents
C1729
Sigma-Aldrich
Cell Neurobiology Techniques
About This Item
ISBN-10:
0-89603-510-7
ISBN-13:
978-0-89603-510-2
UNSPSC Code:
55101509
publication info
A.A. Boulton, et al., ed., Humana Press, 1999, 391 pp., hard cover
manufacturer/tradename
Humana
General description
This book details cutting-edge techniques and readily reproducible methods for establishing neural cell cultures, measuring enzymes and their inhibitors, and using quantitative autoradiography to study monoamine uptake sites and receptors in the brain. Additional methods cover the use of flow cytometry to study developmental neurobiology, applications of magnetic resonance spectroscopy (MRS) to human brain metabolism, and the study of drug metabolism.
Table of Contents
Quantitative Autoradiography of Monoamine Uptake Sites and Receptors in Rat and Mouse Brain,
T Neural Cell Culture Techniques,
Neural Cell Adhesion Molecules.
On the Measurement of Enzymes and their Inhibitors,
Cytochrome P-450 Enzymes: In Vitro Assessment and Clinical Implications.
c-fos Expression as a Marker of Functional Activity in the Brain: Immunohistochemistry.
Immediate-Early Genes as Activity Markers in the CNS. Principles of Drug Metabolism, with an Emphasis on Psychiatric Drugs,
Flow Cytometric Strategies to Study CNS Development. Neurochemistry of Human Postmortem Brain.
Applications of Proton MRS to Study Human Brain Metabolism.
Index.
Choose from one of the most recent versions:
Certificates of Analysis (COA)