Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1497

Sigma-Aldrich

Anti-CCL17/TARC antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-A-152E5.3, Anti-ABCD-2, Anti-SCYA17, Anti-TARC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5-15 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL17(6361)

General description

Chemokine (C-C motif) ligand 17 (CCL17) belonging to the CC chemokine family. It is also known as thymus and activation regulated chemokine (TARC), ABCD-2, SCYA17, A-152E5.3. It is expressed constitutively in thymus and transiently in stimulated peripheral blood mononuclear cells.

Immunogen

purified, E. coli-derived, recombinant human CCL-17.

Application

Anti-CCL17/TARC antibody produced in goat is suitable for the following applications:
  • Immunohistochemistry using formalin-fixed and paraffin-embedded sections at a concentration of 5-15μg/mL.
  • Neutralization
  • Western blotting at a concentration of 0.1-0.2μg/mL

Biochem/physiol Actions

Thymus and activation regulated chemokine (TARC) may play an important role in T cell development as well as in trafficking and activation of mature T cells. It functions as a selective chemo-attractant for T cells that express a class of receptors binding TARC with high affinity and specificity. TARC acts as a functional ligand for CCR4, and CCR4 that are specific receptor for TARC selectively expressed on T cells. In Hodgkin lymphoma (HL) disease it is highly expressed and is a possible biomarker in HL patients. TARC expression is resistant to corticosteroid and is suppressed by PI3K enzyme inhibitors.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maike Sauer et al.
American journal of hematology, 88(2), 113-115 (2012-12-12)
Hodgkin lymphoma (HL) has become one of the best curable cancers. However, better biomarkers are needed for outcome prediction that would allow protecting patients from over- or under-dosing of treatment. Thymus and activation-regulated chemokine/CCL17 (TARC) is highly and specifically elevated
Karl J Staples et al.
The Journal of allergy and clinical immunology, 130(6), 1404-1412 (2012-09-18)
Studies with monocyte-derived macrophages (MDMs) and animal models have suggested a role for alternatively activated (M2) macrophages in asthmatic inflammation, but in vivo evidence for this phenotype in human asthma is lacking. To characterize the phenotype of lung macrophages from asthmatic
T Imai et al.
The Journal of biological chemistry, 271(35), 21514-21521 (1996-08-30)
Precursors of most secreted and cell surface molecules carry signal sequences at their amino termini. Here we describe an efficient signal sequence trap method and isolation of a novel CC chemokine. An expression library was constructed by inserting 5' portion-enriched
T Imai et al.
The Journal of biological chemistry, 272(23), 15036-15042 (1997-06-06)
Thymus and activation-regulated chemokine (TARC) is a recently identified CC chemokine that is expressed constitutively in thymus and transiently in stimulated peripheral blood mononuclear cells. TARC functions as a selective chemoattractant for T cells that express a class of receptors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service