Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1483

Sigma-Aldrich

Anti-Cannabinoid Receptor 2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNR2(1269)
rat ... Cnr2(57302)

Immunogen

fusion protein containing amino acids 1-32 of the rat CB2 receptor

Physical form

Solution in phosphate buffered saline, containing 1 mg/ml bovine serum albumin, 50% glycerol and 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guy A Cabral
Methods in molecular medicine, 123, 41-69 (2006-03-02)
Two cannabinoid receptors have been identified to date. The first of these, designated CB1, is localized primarily in the central nervous system but is also present at lower levels in other tissues. The second receptor, CB2, has been found natively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service