Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1419

Sigma-Aldrich

Cell Dissociation Solution Non-enzymatic 1x

Prepared in Hanks′ balanced salt solution without calcium or magnesium, sterile-filtered, BioReagent, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204

sterility

sterile-filtered

product line

BioReagent

form

solution

concentration

1 ×

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Special non-enzymatic formulation for gently dislodging adherent cell types from plastic or glass surfaces.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankur Jamwal et al.
Metallomics : integrated biometal science, 9(6), 744-756 (2017-05-23)
The present study evaluated the mechanistic underpinnings of the interactive effects of selenium (Se), both inorganic (selenite) and organic (selenomethionine (SeMet)), against arsenite (As-III) cytotoxicity using rainbow trout (Oncorhynchus mykiss) hepatocytes in primary culture. Arsenite is known to induce cytotoxic
Richard Apps et al.
Immunology, 124(3), 322-328 (2008-01-22)
Human leucocyte antigen (HLA)-C is expressed at lower levels than other classical HLA-I molecules on somatic cells. Surface HLA-C proteins can occur as conventionally beta(2)-microglobulin (beta2m)-associated complexes or as open conformers dissociated from peptide and/or beta(2)m. We investigated the conformation
Andrew M Edwards et al.
PloS one, 6(4), e18899-e18899 (2011-04-29)
Staphylococcus aureus is a commensal organism and a frequent cause of skin and soft tissue infections, which can progress to serious invasive disease. This bacterium uses its fibronectin binding proteins (FnBPs) to invade host cells and it has been hypothesised
Ankur Jamwal et al.
Metallomics : integrated biometal science, 8(2), 218-227 (2015-12-18)
The present study evaluated the ameliorative properties of selenium (Se) against cadmium (Cd)-induced oxidative stress, using isolated rainbow trout (Oncorhynchus mykiss) hepatocytes in primary culture as the model experimental system. Cadmium (Cd) is known to induce cytotoxic effects by disrupting
Philip Robinson et al.
PloS one, 6(6), e19802-e19802 (2011-06-23)
Voltage-gated calcium channels (Ca(v)) exist as heteromultimers comprising a pore-forming α(1) with accessory β and α(2)δ subunits which modify channel trafficking and function. We previously showed that α(2)δ-1 (and likely the other mammalian α(2)δ isoforms--α(2)δ-2, 3 and 4) is required

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service