Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-14096

Sigma-Aldrich

Human Mononuclear Cells (hMNC)

Isolated from cord blood, single donor, 1 million cells per pellet

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$177.00

$177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$177.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$177.00


Check Cart for Availability

Request a Bulk Order

biological source

human peripheral blood

packaging

pkg of 1,000,000 cells

morphology

(mononuclear)

application(s)

cell analysis (DNA, RNA, and protein)

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Cell Line Origin

Blood

Application

Mononuclear Cells (MNC) represent the lymphocyte and monocyte fraction isolated from umbilical cord blood or peripheral blood. They are prepared by optimized low-density gradient centrifugation, which effectively reduces the granulocyte content. Red blood cells are gently but thoroughly depleted by proprietary techniques instead of osmotic lysis. This results in ultra-pure mononuclear cells with superior viability and unchanged biological function. In addition, the cells do not clump after thawing, allowing for easy handling and reproducible results.The MNC can be used directly for experiments or as a starting material for the isolation of several cell types, e.g., hematopoietic and endothelial progenitors, monocytes, and lymphocyte subpopulations.

Quality

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Subculture Routine

Click here for more information.

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Ambion, Inc.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Primary cells from PromoCell are available in pelleted form for research applications like gene expression analysis, and cell/protein characterization experiments.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service