Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-14018

Sigma-Aldrich

Human Dermal Blood Endothelial Cells (HDBEC)

Juvenile foreskin, 1 million cells per pellet

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$234.00

$234.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$234.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$234.00


Check Cart for Availability

Request a Bulk Order

biological source

human foreskin (juvenile dermis)

packaging

pkg of 1,000,000 cells

morphology

(endothelial)

application(s)

cell analysis (DNA, RNA, and protein)

relevant disease(s)

cancer; cardiovascular diseases

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Cell Line Origin

Skin

Application

Primary Human Dermal Blood Endothelial Cells (HDBEC) are a subpopulation of the Human Dermal Endothelial Cells. They are isolated from the dermis of juvenile foreskin from a single donor. (Human Dermal Lymphatic Endothelial Cells (HDLEC) from the same donor are available on request.)The cells are routinely analyzed by flow cytometric analysis and immunofluorescent staining: they are >95% CD31 positive and stain positive for von Willebrand factor. In addition, they are >90% negative for podoplanin and stain negative for smooth muscle specific alpha-actin.Blood Endothelial Cells have a key function in physiological processes like vessel tonus, capillary permeability, blood coagulation, fibrolysis, and angiogenesis. They are also useful for disease studies of atherosclerosis, tumor genesis, and thrombosis.

Quality

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Subculture Routine

Click here for more information.

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Ambion, Inc.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary cells from PromoCell are available in pelleted form for research applications like gene expression analysis, and cell/protein characterization experiments.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service