Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-12386

Sigma-Aldrich

Human Uterine Fibroblasts (HUF)

500,000 proliferating cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Product C-12386 is not currently sold in your country. Contact Technical Service

biological source

human uterus (myometrium)

packaging

pkg of 500,000 cells

morphology

(fibroblast)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Uterus

Application

Primary Human Uterine Fibroblasts (HUF) are isolated from the uterine wall and are provided in a proliferating format. During pregnancy, the uterine wall undergoes deep but reversible structural and biochemical changes. The uterine wall is the target of hormones and growth factors that act simultaneously and lead to extensive modifications of the extracellular matrix. This and other morphological and biochemical effects can be studied in vitro using HUF.

Quality

Rigid quality control tests are performed for each lot of Human Fibroblasts. They are tested for cell morphology, adherence rate and cell viability. Flow cytometric analyses of the fibroblast specific antigen CD90 are performed for each lot. Growth performance is tested through multiple passages up to 10 or 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 3500 - 7000 cells per cm2Passage After Thawing: P2Tested Markers: CD90 positiveGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service