Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-12226

Sigma-Aldrich

Human Dermal Blood Endothelial Cells (HDBEC)

Adult, 500,000 proliferating cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Product C-12226 is not currently sold in your country. Contact Technical Service

biological source

human dermis

packaging

pkg of 500,000 cells

morphology

( endothelial)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer; cardiovascular diseases

shipped in

ambient

storage temp.

room temp

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Skin

Application

Primary Human Dermal Blood Endothelial Cells (HDBEC) are a subpopulation of the Human Dermal Endothelial Cells. They are isolated from the dermis of adult skin (different locations) from a single donor and are provided in a proliferating format. (Human Dermal Lymphatic Endothelial Cells (HDLEC) from the same donor are available on request.)The cells are routinely analyzed by flow cytometric analysis and immunofluorescent staining: they are >95% CD31 positive and stain positive for von Willebrand factor. In addition, they are >90% negative for podoplanin and stain negative for smooth muscle specific alpha-actin.Blood Endothelial Cells have a key function in physiological processes like vessel tonus, capillary permeability, blood coagulation, fibrolysis, and angiogenesis. They are also useful for disease studies of atherosclerosis, tumor genesis, and thrombosis.

Quality

Rigid quality control tests are performed for each lot of Microvascular Endothelial Cells. They are tested for cell morphology and cell type specific markers, e.g. von Willebrand Factor (vWF), CD31 and Podoplanin using flow cytometric analyses. Growth performance is tested through multiple passages up to 15 population doublings (PD) without antibiotics or antimycotics. In addition, all cells have been tested forthe absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 10000 - 20000 cells per cm2Passage After Thawing: P2Tested Markers: Podoplanin negative, CD31 positiveGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Related Content

Cell culture protocol: the endothelial tube formation assay to study angiogenesis using HUVECs and other endothelial cell types. Explore over 350 PromoCell products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service