Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C11308-HH

Sigma-Aldrich

DMT-dC(ac) Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):

DMT-dC(ac) Amidite, N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N4-Acetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H50N5O8P
CAS Number:
Molecular Weight:
771.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

assay

≥98% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

packaging

pkg of 12 × 1 g C113081-12
pkg of 12 × 2 g C113082-12

technique(s)

oligo synthesis: suitable

color

white to off-white

λ

conforms (UV/VIS Identity)

compatibility

configured for PerkinElmer
configured for Polygen

nucleoside profile

base: cytidine
base protecting group: acetyl
5' protecting group: DMT
deprotection: fast

Storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(C)=O)=NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C41H50N5O8P/c1-28(2)46(29(3)4)55(52-25-11-23-42)54-36-26-39(45-24-22-38(43-30(5)47)44-40(45)48)53-37(36)27-51-41(31-12-9-8-10-13-31,32-14-18-34(49-6)19-15-32)33-16-20-35(50-7)21-17-33/h8-10,12-22,24,28-29,36-37,39H,11,25-27H2,1-7H3,(H,43,44,47,48)/t36-,37+,39+,55?/m0/s1

Inchi Key

MECWEBCHRKVTNV-RLWDVSNXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service