Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1041

Sigma-Aldrich

Monoclonal Anti-Desmosomal Cytokeratin antibody produced in mouse

clone DK80.20, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DK80.20, monoclonal

contains

15 mM sodium azide

species reactivity

bovine, human

technique(s)

electron microscopy: suitable
immunohistochemistry (frozen sections): suitable
indirect immunofluorescence: 1:100 using frozen bovine tissue sections
western blot: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT8(3856)

Specificity

Reacts with cytokeratin peptide 8. The antibody detects desmosomes in frozen sections of bovine and human, non-stratified epithelium.

Immunogen

cytoskeletal proteins from the bovine mammary gland epithelial cell line BMGE

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service