Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1038

Sigma-Aldrich

Complement C4 deficient serum from guinea pig

for complement assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

biological source

guinea pig

Quality Level

form

solution

technique(s)

activity assay: suitable

shipped in

dry ice

storage temp.

−70°C

Application

Complement C4 deficiency has long been associated with the disease systemic lupus erythematosus (SLE) and severe kidney disorders. Screening of the mutations which lead to C4 deficiency may assist in studies of these diseases and disorders. It has been demonstrated that 40-60% of patients with SLE contained either heterozygous or homozygous deficiencies in C4 compliment components via C4A and C4B genes. In particular, research has shown that complete C4A and C4B deficiencies occur through deleterious mutations at exon 13 and within a 2.6-kb genomic region spanning exons 20-29.

Analysis Note

C4 is deficient as judged by a highly sensitive hemolytic assay and Ouchterlony immunodiffusion method.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complete complement components C4A and C4B deficiencies in human kidney diseases and systemic lupus erythematosus.
Yang Y., et al.
Journal of Immunology, 15, 2803-2814 (2004)
Manfred Nairz et al.
Frontiers in cellular and infection microbiology, 7, 110-110 (2017-04-27)
Genetic and dietary forms of iron overload have distinctive clinical and pathophysiological features. HFE-associated hereditary hemochromatosis is characterized by overwhelming intestinal iron absorption, parenchymal iron deposition, and macrophage iron depletion. In contrast, excessive dietary iron intake results in iron deposition
Anamarija Markota et al.
PloS one, 14(7), e0218332-e0218332 (2019-07-06)
Clinical observations in inflammatory bowel disease patients and experimental studies in rodents suggest that iron in the intestinal lumen derived from iron-rich food or oral iron supplementation could exacerbate inflammation and that iron depletion from the diet could be protective.
Wannaporn Ittiprasert et al.
Journal of autoimmunity, 25(1), 77-84 (2005-07-07)
The complement component C4 is encoded by two genes: C4A and C4B on human chromosome 6p in the major histocompatibility complex (MHC). Most studies have linked the deficiencies in C4 with systemic lupus erythematosus (SLE) in Angio-Irish, North American, Black

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service