Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0922

Sigma-Aldrich

Tyr-Corticotropin Releasing Factor Ovine

≥97% (HPLC), Synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C214H348N60O65S
CAS Number:
Molecular Weight:
4833.48
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Ser-Gln-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Thr-Lys-Ala-Asp-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-Leu-Leu-Asp-Ile-Ala-NH2

Application

CRF analog used to prepare iodinated CRF for receptor studies and immunoassay

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding of [125I]Tyr-corticotropin-releasing factor to rabbit aorta is reduced by removal of the endothelium.
M R Dashwood et al.
European journal of pharmacology, 135(1), 111-112 (1987-03-03)
I Q Assil et al.
Peptides, 22(7), 1055-1061 (2001-07-11)
The methionine residues in Tyr-corticotropin-releasing factor (CRF) and Tyr-sauvagine radioligands are subject to oxidation, which renders them biologically inactive. Therefore [Tyr(0,) Gln(1,) Leu(17)]sauvagine (YQLS), in which the methionine was replaced with leucine was synthesized and labeled with (125)Iodine using chloramine-T.
A V Turnbull et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 215(1), 1-10 (1997-05-01)
Corticotropin-releasing factor (CRF) is a 41-amino acid neuropeptide, which is recognized as a critical mediator of complimentary, stress-related endocrine, autonomic, and behavioral responses in mammalian species. CRF belongs to a family of structurally related peptides including frogskin sauvagine and fish
K Eckart et al.
Current medicinal chemistry, 6(11), 1035-1053 (1999-10-16)
Corticotropin-releasing factor (CRF), urocortin, sauvagine and urotensin I form the CRF family. These peptides bind with different affinities to two subtypes of CRF receptor (CRFR), CRFR1 and CRFR2. The latter exists as two splice variants, the neuronal CRFR2a and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service