Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0864

Sigma-Aldrich

Anti-phospho-Cortactin (pTyr466) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80-85 kDa

species reactivity

mouse

technique(s)

western blot: 0.1-1.0 μg/mL using NIH3T3 cells expressing activated c-Src

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pTyr466)

Gene Information

mouse ... Cttn(13043)

General description

Cortactin is encoded by the CTTN gene on chromosome 11 in humans. It is a cytoskeleton protein that facilitates assembly of cortical actin. It is expressed in most adherent cells and is a prominent substrate of protein-tyrosine kinase Src.

Immunogen

synthetic phosphopeptide derived from the region of mouse cortactin that is phosphorylated on tyrosine 466.

Application

Anti-phospho-Cortactin (pTyr466) antibody produced in rabbit is suitable for western blotting at a concentration of 0.1-1.0μg/mL using NIH3T3 cells expressing activated c-Src.

Biochem/physiol Actions

Cortactin is activated by phosphorylation of tyrosine kinases or serine/threonine kinases. It plays an important role in promoting lamellipodia formation, invadopodia formation, cell migration, and endocytosis. Cortactin regulates the interactions between components of adherens-type junctions and helps in organizing the cytoskeleton, cell adhesion structures of epithelia and carcinoma cells. During apoptosis, the encoded protein is degraded in a caspase-dependent manner. It acts as a regulator in invasion and migration of tumor cells. The overexpression of cortactin causes osteosarcoma carcinogenesis and along with FAK increases the risk of oral premalignant lesions.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, containing 1.0 mg/ml BSA (IgG, protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Zhao et al.
World journal of surgical oncology, 11, 74-74 (2013-03-23)
Cortactin is an important regulator involved in invasion and migration of hepatocellular carcinoma (HCC). The aim of this study was to elucidate the forecasting role of cortactin in resectable HCCs. We compared the invasiveness and motility among liver epithelial cell
Cecilia Folio et al.
Cancer biomarkers : section A of Disease markers, 10(1), 35-41 (2012-02-03)
The cortactin (CTTN) gene has been found, by transcriptomic profiling, to be overexpressed in pediatric osteosarcoma. The location of CTTN at 11q13 and the role of cortactin in cytoskeleton restructuring make CTTN of interest as a potential biomarker for osteosarcoma.
Roger J Daly
The Biochemical journal, 382(Pt 1), 13-25 (2004-06-10)
Cortactin was first identified over a decade ago, and its initial characterization as both an F-actin binding protein and v-Src substrate suggested that it was likely to be a key regulator of actin rearrangements in response to tyrosine kinase signalling.
Kellye C Kirkbride et al.
Cytoskeleton (Hoboken, N.J.), 69(9), 625-643 (2012-09-20)
Cortactin is a branched actin regulator and tumor-overexpressed protein that promotes vesicular trafficking at a variety of cellular sites, including endosomes and the trans-Golgi network. To better understand its role in secretory trafficking, we investigated its function in Golgi homeostasis.
Aviv Bergman et al.
Cell adhesion & migration, 8(3), 273-279 (2014-04-10)
Invadopodia are dynamic protrusions in motile tumor cells whose function is to degrade extracellular matrix so that cells can enter into new environments. Invadopodia are specifically identified by microscopy as proteolytic invasive protrusions containing TKS5 and cortactin. The increasing complexity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service