C0853
Anti-Crk-II antibody produced in rabbit
IgG fraction of antiserum, buffered aqueous solution
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 38 kDa
species reactivity
human, mouse, chicken
technique(s)
microarray: suitable
western blot: 1:3,000 using whole extract of the Burkitt lymphoma Raji cell line
western blot: 1:5,000 using whole extract of chicken fibroblasts
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... CRK(1398)
mouse ... Crk(12928)
General description
Anti-Crk-II specifically recognizes human and chicken Crk-II (38 kDa).
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service