Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0809

Sigma-Aldrich

Chorionic gonadotropin human

from human pregnancy urine, lyophilized powder, vial of 1 mg

Synonym(s):

Choriogonin, HCG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

biological source

human pregnancy urine

sterility

sterile-filtered

form

lyophilized powder

mol wt

37.9 kDa

packaging

vial of 1 mg

drug control

Home Office Schedule 4.2

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CGB5(93659)

Looking for similar products? Visit Product Comparison Guide

General description

Glycoprotein hormone consisting of a 92-amino acid α-chain which is identical to that of LH, FSH, and TSH; and a distinct 149-amino acid β-chain. Produced by chorionic tissue and responsible for maintaining the corpus luteum during early pregnancy. Also produced by trophoblast cell neoplasms, which are marked by elevated free hCG β-chains and hyperglycosylated hCG.

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Repr. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bridget A Quinn et al.
PloS one, 4(12), e8404-e8404 (2010-01-05)
Approximately one out of every ten cases of epithelial ovarian cancer (EOC) is inherited. The majority of inherited cases of EOC result from mutations in the breast cancer associated gene 1 (BRCA1). In addition to mutation of BRCA1, mutation of
Corine K Lau et al.
Molecular biology of the cell, 20(18), 4043-4058 (2009-07-31)
Mitosis in higher eukaryotes is marked by the sequential assembly of two massive structures: the mitotic spindle and the nucleus. Nuclear assembly itself requires the precise formation of both nuclear membranes and nuclear pore complexes. Previously, importin alpha/beta and RanGTP
Mary E Sabatini et al.
Journal of ovarian research, 4(1), 8-8 (2011-05-25)
Metformin, an oral biguanide traditionally used for the treatment of type 2 diabetes, is widely used for the management of polycystic ovary syndrome (PCOS)-related anovulation. Because of the significant prevalence of insulin resistance and glucose intolerance in PCOS patients, and
Justine Mathieu-Denoncourt et al.
General and comparative endocrinology, 219, 53-63 (2015-02-11)
Polymer flexibility and elasticity is enhanced by plasticizers. However, plasticizers are often not covalently bound to plastics and thus can leach from products into the environment. Much research effort has focused on their effects in mammalian species, but data on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service