Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0646

Sigma-Aldrich

Cytodex® 1 microcarrier beads

Cytodex 1, 60-87 μm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41121800
NACRES:
NB.22

material

dextran beads

sterility

non-sterile

particle size

60-87 μm

density

1.03 g/cm3 at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

A general-purpose microcarrier for most established, anchorage-dependent cell lines.

High yield culture of anchorage-dependent cells: Microcarrier culture is a technique which makes possible the practical high yield culture of anchorage-dependent cells. Cytodex has been specifically developed for the culture of a wide range of animal cells in culture volumes ranging from a few mL to more than 6000 L. Using Cytodex in simple suspension culture systems provides yields of several million cells per mL.

Suitable for most established cell lines: Formed by substituting a cross-linked dextran matrix with positively charged DEAE* groups distributed throughout the matrix, is a general purpose microcarrier. It is particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains.

Application

For general microcarrier culture.

The particle size of this product averages 190um when swelled in liquid solutions (as in use). The particle size of 60-87 um relates to the beads in dry form.

Features and Benefits

  • General-purpose microcarrier for most established, anchorage-dependent cell lines.
  • Contains positive charges throughout the microporous matrix.
  • Transparent for easy microscopic examination of attached cells.

Other Notes

Dextran beads with positive-charged DEAE groups throughout matrix.

Preparation Note

Prior to use, swell beads in PBS (50 mL/g) and autoclave at 120 °C.

Legal Information

Cytodex is a registered trademark of Cytiva

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quanyu Zhao et al.
Biotechnology progress, 19(5), 1569-1573 (2003-10-04)
Toward the development of an in vitro cultivation of marine sponge cells for sustainable production of bioactive metabolites, the attachment characteristics of marine sponge cells of Hymeniacidon perleve on three types of microcarriers, Hillex, Cytodex 3, and glass beads, were
Denver C Surrao et al.
Tissue engineering. Part A, 17(15-16), 1959-1967 (2011-04-01)
Tissue engineering is a promising approach for articular cartilage repair; however, it is challenging to produce adequate amounts of tissue in vitro from the limited number of cells that can be extracted from an individual. Relatively few cell expansion methods
Pierre Danhier et al.
PloS one, 8(1), e53324-e53324 (2013-02-06)
Cell detachment is a procedure routinely performed in cell culture and a necessary step in many biochemical assays including the determination of oxygen consumption rates (OCR) in vitro. In vivo, cell detachment has been shown to exert profound metabolic influences
S Mehdi Hassanzadeh et al.
Pakistan journal of biological sciences : PJBS, 14(7), 441-448 (2011-09-10)
To achieve higher titer of rabies virus higher density of host cells will need. In this study, capability of FibraCel disks packed in 500 mL spinner basket versus Cytodex-1 in 500 mL spinner flask was investigated for propagation of Vero
Christopher J Hewitt et al.
Biotechnology letters, 33(11), 2325-2335 (2011-07-20)
The effects on human mesenchymal stem cell growth of choosing either of two spinner flask impeller geometries, two microcarrier concentrations and two cell concentrations (seeding densities) were investigated. Cytodex 3 microcarriers were not damaged when held at the minimum speed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service