C0224
Monoclonal Anti-Cdc6 antibody produced in mouse
clone DCS-180, ascites fluid
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
DCS-180, monoclonal
mol wt
antigen 62 kDa
contains
15 mM sodium azide
species reactivity
human
technique(s)
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1:2,000 using a HeLa cell extract
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... CDC6(990)
Immunogen
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service