BR36680
BRAND® BLAUBRAND® volumetric flask with serialized numbers, glass stopper
capacity 5 mL, joint: ST/NS 10/16, polypropylene stopper, accuracy: 0.04 mL, clear glass
About This Item
material
clear glass
polypropylene stopper
wide-neck
joint
joint: ST/NS10/16
packaging
pkg of 12 ea
manufacturer/tradename
BRAND 27452
measuring range
accuracy: 0.04 mL
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service