Key Documents
BR36476
BRAND® BLAUBRAND® volumetric flask, trapezoidal, with serialized numbers
capacity 2 mL, joint: ST/NS 7/16, polypropylene stopper
About This Item
UNSPSC Code:
41121804
Pricing and availability is not currently available.
material
polypropylene stopper
joint
joint: ST/NS7/16
packaging
pkg of 12 ea
manufacturer/tradename
BRAND 27452
capacity
2 mL
General description
Borosilicate 3.3. To DIN EN ISO 1042. Calibrated to contain (TC, In).All BLAUBRAND(R) volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certi
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)