Key Documents
BR29955
BRAND® BLAUBRAND® bulb pipette with serialized numbers, calibrated to deliver (TD, EX)
capacity 3 mL, one mark
About This Item
UNSPSC Code:
41121510
Pricing and availability is not currently available.
material
AR-Glass
feature
supplementary to DIN range
packaging
pkg of 12 ea
manufacturer/tradename
BRAND 27452
capacity
3 mL
General description
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.According to DIN EN ISO 648.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)