Key Documents
BR27452
BRAND® BLAUBRAND® graduated pipette with serialized numbers, calibrated to contain
capacity 0.1 mL
About This Item
UNSPSC Code:
41121500
Pricing and availability is not currently available.
material
AR-Glass
feature
graduation
supplementary to DIN range
packaging
pkg of 12 ea
manufacturer/tradename
BRAND 27452
L
360 mm
capacity
0.1 mL
General description
Boro 3.3. DIN EN ISO 1042. Calibrated to contain (TC, In).All BLAUBRAND® volumetric flasks are delivered with a batch number and batch certificate included in the original packaging.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)