shipped in
wet ice
Storage temp.
−20°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Guard et al.
European journal of pharmacology, 240(2-3), 177-184 (1993-08-24)
The pharmacological profile of [125I][Tyr4]bombesin binding to gastrin-releasing peptide- and neuromedin B-preferring sites has been investigated in rat cerebral cortex and olfactory bulb membranes, respectively. [125I][Tyr4]bombesin specific binding to cerebral cortex membranes was displayed biphasically by gastrin releasing peptide and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service