Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BOCCCK

Sigma-Aldrich

Cholecystokinin Fragment Kit, Boc Protected

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

wet ice

storage temp.

−20°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nikolaus et al.
Neural plasticity, 6(4), 77-89 (2000-03-14)
The neuropeptide substance P is known to have mnemogenic and reinforcing actions and can exert neurotrophic and regenerative effects in vitro as well as in vivo. Furthermore, our previous work in the rat showed that either pre- or post-lesion treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service