Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0032

Sigma-Aldrich

BMY 45778

≥98% (HPLC)

Synonym(s):

2-[3-[4-(4,5-diphenyl-2-oxazolyl)-5-oxazolyl]phenoxy]acetic acid, BMY-045778, BMY45778

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18N2O5
CAS Number:
Molecular Weight:
438.43
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

InChI

1S/C26H18N2O5/c29-21(30)15-31-20-13-7-12-19(14-20)24-23(27-16-32-24)26-28-22(17-8-3-1-4-9-17)25(33-26)18-10-5-2-6-11-18/h1-14,16H,15H2,(H,29,30)

InChI key

DSRSEEYZGWTODH-UHFFFAOYSA-N

Biochem/physiol Actions

BMY 45778 is a non-prostanoid prostacyclin agonist.

Features and Benefits

BMY 45778 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service