Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0022

Sigma-Aldrich

BMS-433771 bis-hydrochloride monohydrate

≥98% (HPLC)

Synonym(s):

1-Cyclopropyl-3-[[1-(4-hydroxybutyl)-1H-benzimidazol-2-yl]methyl]-1,3-dihydroimidazo[4,5-c]pyridin-2-one bis-hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N5O2 · 2HCl · H2O
CAS Number:
Molecular Weight:
468.38
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[H]O[H].[H]Cl.[H]Cl.OCCCCN1C2=CC=CC=C2N=C1CN3C(C=NC=C4)=C4N(C5CC5)C3=O

InChI

1S/C21H23N5O2.2ClH.H2O/c27-12-4-3-11-24-17-6-2-1-5-16(17)23-20(24)14-25-19-13-22-10-9-18(19)26(21(25)28)15-7-8-15;;;/h1-2,5-6,9-10,13,15,27H,3-4,7-8,11-12,14H2;2*1H;1H2

InChI key

DMSGBVAHVFUZMJ-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-433771 bis-hydrochloride monohydrate is a respiratory syncytial virus (RSV) fusion inhibitor with an EC50 value of 10-12 nM. BMS-433771 bis-hydrochloride targets the RSV fusion (RSVF) protein, preventing viral entry. BMS-433771 bis-hydrochloride is orally available.
BMS-433771 bis-hydrochloride monohydrate is a respiratory syncytial virus (RSV) fusion inhibitor.

Features and Benefits

BMS-433771 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service