Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0020

Sigma-Aldrich

BMS-646786

≥98% (HPLC)

Synonym(s):

BPTU, N-[2-[2-(1,1-Dimethylethyl)phenoxy]-3-pyridinyl]-N′-[4-(trifluoromethoxy)phenyl]-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22F3N3O3
CAS Number:
Molecular Weight:
445.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(NC1=CC=C(OC(F)(F)F)C=C1)NC2=C(OC3=CC=CC=C3C(C)(C)C)N=CC=C2

InChI

1S/C23H22F3N3O3/c1-22(2,3)17-7-4-5-9-19(17)31-20-18(8-6-14-27-20)29-21(30)28-15-10-12-16(13-11-15)32-23(24,25)26/h4-14H,1-3H3,(H2,28,29,30)

InChI key

AHFLGPTXSIRAQK-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-646786 is a potent and specific inhibitor of P2Y1 purinergic receptor that inhibits ADP-mediated platelet aggregation in human blood samples. BMS-646786 significantly reduces thrombus weight in a rat arterial thrombosis model with a limited effect on bleeding.
BMS-646786 is a potent and specific inhibitor of P2Y1 purinergic receptor.

Features and Benefits

BMS-646786 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiao-Hsuan Chao et al.
PLoS pathogens, 15(4), e1007625-e1007625 (2019-04-23)
Dengue virus (DENV) infection, the most common mosquito-transmitted viral infection, can cause a range of diseases from self-limiting dengue fever to life-threatening dengue hemorrhagic fever and shock syndrome. Thrombocytopenia is a major characteristic observed in both mild and severe dengue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service