Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0001

Sigma-Aldrich

BMS-341400 methanesulfonate

≥98% (HPLC)

Synonym(s):

1-[9-[[(3-Chloro-4-methoxyphenyl)methyl]amino]-3-ethyl-3H-pyrazolo[4′,3′:5,6]pyrido[3,4-d]pyridazin-6-yl]-N-methyl-H-imidazole-4-carboxamide methanesulfonate (1:1), 1-[9-[[(3-Chloro-4-methoxyphenyl)methyl]amino]-3-ethyl-3Hpyrazolo[4′,3′:5,6]pyrido[3,4-d]pyridazin-6-yl]-4-methylimidazolecarboxamide methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClN9O2 · CH4O3S
CAS Number:
Molecular Weight:
588.04
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(C)(O)=O.ClC1=CC(CNC2=C(C(C=NN3CC)=C3N=C4)C4=C(N5C=NC(C(NC)=O)=C5)N=N2)=CC=C1OC

InChI

1S/C23H22ClN9O2.CH4O3S/c1-4-33-21-15(10-29-33)19-14(9-27-21)22(32-11-17(28-12-32)23(34)25-2)31-30-20(19)26-8-13-5-6-18(35-3)16(24)7-13;1-5(2,3)4/h5-7,9-12H,4,8H2,1-3H3,(H,25,34)(H,26,30);1H3,(H,2,3,4)

InChI key

NTAGDQYHYAUFMS-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-341400 is a selective and potent phosphodiesterase 5 (PDE 5) inhibitor.
BMS-341400 is a selective and potent phosphodiesterase 5 (PDE 5) inhibitor. BMS-341400 has an IC50 value of 0.3 nM and is 150-fold selective for PDE5 over PDE6 and over 10,000-fold selective for PDE5 over PDE1, PDE2, PDE3, and PDE4.

Features and Benefits

BMS-341400 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service