BLD9601
GenElute™ 96 Well Blood Genomic DNA Purification Kit
1 x 96 well plate, sufficient for 1 96-well plate purifications
Synonym(s):
High Throughput Purification, Blood Genomic DNA
About This Item
Recommended Products
usage
sufficient for 1 96-well plate purifications
quality
1 x 96 well plate
Storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
General description
The starting materials are lysed at room temperature in a lysis buffer which contains chaotropic salts in the presence of Proteinase K. The addition of ethanol ensures the appropriate conditions to bind the DNA to the silica membrane of the Blood Binding Plate. After washing the bound DNA to remove contaminants, the pure genomic DNA is eluted in the Blood Elution Solution included in the kit.
The purified genomic DNA is ready for use in downstream applications such as PCR, Southern blotting, or any kind of enzymatic reaction.
Application
- PCR
- restriction digest
- Southern blots
- cloning
- enzymatic reactions
Features and Benefits
- Starting material: 200 μl fresh or frozen blood
- Expected yield: 4 - 6 μg/well
- Elution volume: 50 - 200 μl
- Time required: 70 min/plate
- Innovative wash plate minimizes risk of cross-contamination
- Reduced risk of clogging
- Complete processing possible at room temperature
Principle
Other Notes
Legal Information
recommended
related product
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system, Respiratory system
Storage Class
3 - Flammable liquids
flash_point_f
73.4 °F
flash_point_c
23 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service