Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

BCS-572

CoolBox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103000
NACRES:
NB.22

feature

Includes CoolBox XT System and CoolRack XT M-PCR (holds 12 x 1.5-2.0 mL microcentrifuge tubes and 6 rows of PCR strips.)

manufacturer/tradename

BioCision BCS-572

external L × W × H

20.0 × 16.0 × 15.0 cm

color

green

Looking for similar products? Visit Product Comparison Guide

General description

Includes CoolBox XT System, automation-compatible CoolRack XT M-PCR to hold 12 x 1.5 or 2.0ml microcentrifuge tubes and 6 rows of PCR strip tubes.

Easily transfer samples from the CoolBox XT cooling workstation to the robotic platform for brief cold processing (approx 20-30 minutes <10°C). Ideal for any type of PCR reaction- from simple amplification to preparing RNA to preparing q-PCR, dPCR, or ddPCR reactions.

Applications include qPCR, dPCR, ddPCR, ampification for cloning and DNA sequencing.

Legal Information

CoolBox is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service